Learn More
3M™ Maintenance Sorbent Multi-Format
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W):
12 x 1520 cm
missing translation for 'unitSize'
Pack of 3
Specifications
Specifications
| Type | Maintenance Sorbent, Multi-Format |
| Packaging Quantity | 3/Cs. |
| Dimensions (L x W) | 12 x 1520 cm |
| Weight (Metric) | 8.2 kg |