missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid, Reagent Grade, 99.5+%, Thermo Scientific™

43.30€ - 155.00€

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3H4O4
Molecular Weight (g/mol) 104.061
MDL Number MFCD00002707
InChI Key OFOBLEOULBTSOW-UHFFFAOYSA-N
Synonym malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
PubChem CID 867
ChEBI CHEBI:30794
IUPAC Name propanedioic acid
SMILES C(C(=O)O)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15421247
View Documents
Thermo Scientific Alfa Aesar
031715.22
100g
43.30€
100g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15431247
View Documents
Thermo Scientific Alfa Aesar
031715.36
500g
155.00€
500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

141-82-2
104.061
OFOBLEOULBTSOW-UHFFFAOYSA-N
867
propanedioic acid
C3H4O4
MFCD00002707
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
CHEBI:30794
C(C(=O)O)C(=O)O

Specifications

141-82-2
157°C (314°F)
H2C(CO2H)2
1751370
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
OFOBLEOULBTSOW-UHFFFAOYSA-N
propanedioic acid
867
104.06
Reagent
1.62 g/mL
C3H4O4
MFCD00002707
14,5710
Soluble in water,sodium hydroxide and alcohol; Slightly soluble in pyridine and ether.
C(C(=O)O)C(=O)O
104.061
CHEBI:30794
≥99.5%
Malonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H318-H335

missing translation for 'einecsNumber' : 205-503-0

missing translation for 'rtecsNumber' : OO8175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.