Learn More
Thermo Scientific™ Malt Extract Agar
Description
Detect, isolate and enumerate yeasts and molds with ready-to-use Thermo Scientific™ Oxoid™ Malt Extract Agar. Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation. Due to the acidic nature of the medium, it inhibits the growth of bacteria resulting in improved isolation of yeasts and molds from materials heavily infected with bacteria.
Use Malt Extract Agar for detection, isolation and enumeration of yeasts and molds.
- Supports luxuriant growth of yeasts and molds while inhibiting the growth of bacteria.
Ready-to-use convenience of prepared media. Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Ocher yellow, transparent |
| Content And Storage | 2 to 25°C |
| Description | Malt Extract Agar |
| Format | 90mm Plate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 17g ± 5 % |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |