Learn More
Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated)
Product Code. p-200112232
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. 10321063
Brand: Thermo Scientific™ CM0059B
Detect, isolate and enumerate yeasts and molds with typical morphology and pigmentation using medium.
Detect, isolate and enumerate yeasts and molds with Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated). Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation.
Malt Extract Agar is similar to the formula of Galloway and Burgess1 used for the detection, isolation, and enumeration of yeasts and molds. For mycological counts, it may be desirable to prepare the more acid medium in order to suppress bacterial growth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
Description Malt Extract Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.0L medium
compliance-icons
Product Identifier
  • MALT EXCTRACT AGAR
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Barley, malt
For Laboratory Use Only