Learn More
Abeba™ Manager 3120 Shoes
All calf leather construction. ESD dissipative.
Brand: Abeba™ 3120/40
Additional Details : Weight : 1.20000kg
Description
- CE
- Calf leather
- Black
- Antistatic leather outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Leather insole
Specifications
Black | |
Leather | |
Leather |
Microfiber | |
40 | |
3120 |