Learn More
Abeba™ Manager 3121 Shoes
All calf leather construction. ESD dissipative.
Brand: Abeba™ 3121/41
Additional Details : Weight : 1.20000kg
Description
- CE
- Calf leather
- Brown
- Antistatic leather outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Leather insole
Specifications
Brown | |
Leather | |
Leather |
Microfiber | |
41 | |
3121 |