Learn More
Abeba™ Manager 3121 Shoes
All calf leather construction. ESD dissipative.
Brand:  Abeba™ 3121/41
Additional Details : Weight : 1.20000kg
 View more versions of this product
Product Code. 15628787
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE
  • Calf leather
  • Brown
  • Antistatic leather outsole sole
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Leather insole
Specifications
Specifications
Brown
Leather
Leather
Microfiber
41
3121