Learn More
VACUUBRAND™ Manometers
Click to view available options
Pressure Range:
0 to 6 bar
1 to 1000 mbar
missing translation for 'unitSize'
Each
Specifications
Specifications
| For Use With (Application) | For vacuum adjustment device |
| For Use With (Equipment) | For vacuum adjustment device |
| Pressure Range | 1 to 1000 mbar |
| Product Type | Manometer |