Learn More
Macherey-Nagel™ Manual Crimper
240.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10003061
|
Macherey-Nagel™
735113 |
13 mm Aluminum Crimp Caps |
240.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
10419561
|
Macherey-Nagel™
735126 |
8 mm Aluminum Caps |
240.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
10216911
|
Macherey-Nagel™
735120 |
20 mm Aluminum Crimp Caps |
240.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
10246811
|
Macherey-Nagel™
735111 |
11 mm Aluminum Crimp Caps |
240.00€
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Specifications
Manual Crimper | |
1 Each |