Learn More
Thermo Scientific™ Manual Crimpers
Product Code. 12490078
Change view
Click to view available options
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
12490078 Each
12410268 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12490078 Supplier Thermo Scientific™ Supplier No. C4012103
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Achieve secure, repeatable crimps or remove seals with minimized risk of damage with Thermo Scientific manual crimpers.
  • Lightweight, precision crimpers attach seals to chromatography or serum vials.
  • Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals.
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness.
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 11 mm
For Use With (Equipment) Autosampler Vial Closures
Product Type Manual Crimper