Learn More
Invitrogen™ Human MAP4K5 (KHS1), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q9Y4K4 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 11183 |
| Molecular Weight (g/mol) | 122.5 kDa |
| Name | Human MAP4K5 (KHS1), GST Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | 4432415E19Rik; fl74c10; GCKR; germinal center kinase-related; KHS; KHS1; Kinase homologous to SPS1/STE20; LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 5; map4k5; MAPK/ERK kinase kinase kinase 5; MAPKKKK5; MEK kinase kinase 5; MEKKK 5; mitogen-activated protein kinase kinase kinase kinase 5; RGD1562028; wu:fl74c10; zgc:55719; zgc:55985 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.