missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein
Brand: Invitrogen PV3304
Additional Details : Weight : 3.36000kg
Description
MAPK14 (p38 alpha) is a member of the stress-activated protein kinase class of MAPKs.Specifications
NP_620581 | |
1432 | |
10μg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
mitogen-activated protein kinase 14 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
67.6 kDa | |
MAPK14 (p38 α) | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
MAPK14 | |
E. coli | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.