Learn More
Maprotiline hydrochloride, 99%, Thermo Scientific™
Selective norepinephrine uptake inhibitor
94.31€ - 294.00€
Chemical Identifiers
| CAS | 10347-81-6 |
|---|---|
| Molecular Formula | C20H24ClN |
| Molecular Weight (g/mol) | 313.87 |
| MDL Number | MFCD00079464 |
| InChI Key | NZDMFGKECODQRY-UHFFFAOYSA-N |
| Synonym | maprotiline hydrochloride, maprotiline hcl, ludiomil, psymion, maprotilline hcl, ciba 34276 ba, unii-7c8j54pvfi, deprilept, 9-gamma-methylaminopropyl-9,10-dihydro-9,10-ethanoanthracene hydrochloride |
| PubChem CID | 71478 |
| IUPAC Name | hydrogen methyl(3-{tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-1-yl}propyl)amine chloride |
| SMILES | [H+].[Cl-].CNCCCC12CCC(C3=CC=CC=C13)C1=CC=CC=C21 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15414729
|
Thermo Scientific Alfa Aesar
J62321.03 |
1g |
96.20€ 94.31€ / 1g Save 1.89€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15424729
|
Thermo Scientific Alfa Aesar
J62321.06 |
5g |
300.00€ 294.00€ / 5g Save 6.00€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMaprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.
Solubility
Soluble to 50mg/ml in water. Soluble in ethanol at 10mM
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| 10347-81-6 | |
| 313.87 | |
| NZDMFGKECODQRY-UHFFFAOYSA-N | |
| 71478 | |
| [H+].[Cl-].CNCCCC12CCC(C3=CC=CC=C13)C1=CC=CC=C21 |
| C20H24ClN | |
| MFCD00079464 | |
| maprotiline hydrochloride, maprotiline hcl, ludiomil, psymion, maprotilline hcl, ciba 34276 ba, unii-7c8j54pvfi, deprilept, 9-gamma-methylaminopropyl-9,10-dihydro-9,10-ethanoanthracene hydrochloride | |
| hydrogen methyl(3-{tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-1-yl}propyl)amine chloride |
Specifications
| 10347-81-6 | |
| MFCD00079464 | |
| 14,5748 | |
| Soluble to 50mg/ml in water; Soluble in ethanol at 10mM | |
| [H+].[Cl-].CNCCCC12CCC(C3=CC=CC=C13)C1=CC=CC=C21 | |
| 313.87 | |
| 313.87 | |
| Maprotiline hydrochloride |
| C20H24ClN | |
| 5463242 | |
| maprotiline hydrochloride, maprotiline hcl, ludiomil, psymion, maprotilline hcl, ciba 34276 ba, unii-7c8j54pvfi, deprilept, 9-gamma-methylaminopropyl-9,10-dihydro-9,10-ethanoanthracene hydrochloride | |
| NZDMFGKECODQRY-UHFFFAOYSA-N | |
| hydrogen methyl(3-{tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-1-yl}propyl)amine chloride | |
| 71478 | |
| 99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 233-758-8
missing translation for 'rtecsNumber' : KJ4555000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.