Learn More
Emma Safety Footwear Marco (Frontier 112) Safety Shoes
Made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
Brand:  Emma Safety Footwear MM11209043
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 15865771
  • 210.00€ / 1 pair
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Thermoplastic Polyurethane Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver & Wellness leather
43
Laces
Marco (Frontier 112)
Leather
Steel