Learn More
Thermo Scientific™ Margatoxin, 99+%
Product Code. 15433759
Click to view available options
Quantity:
0.1 mg
5 x 10 μg
5 x 5 μg
Unit Size:
0.1mg
Each
Product Code. 15433759
Brand: Thermo Scientific™ J61103.!Z
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Description White
Quantity 0.1 mg
Assay 99+%
CAS 145808-47-5
Molecular Formula C178H286N52O50S7
Formula Weight 4178.95
Synonym rMargatoxin; MgTx
Product Type Margatoxin
Content And Storage Ambient temperatures
For Use With (Application) Peptide that blocks voltage-dependent Kv1.3 potassium channels
Form Powder