missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
GREENER_CHOICE

Product Code. 10746231 Shop All Thermo Scientific Products
Click to view available options
Size Range:
100 to 1000 bp
1500 to 10000 bp
80 to 10000 bp
80 to 1031 bp
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 10746231

Brand: Thermo Scientific™ SM1283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.

Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.

Highlights

  • Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Bromophenol Blue
Product Type DNA Ladder Mix
Ready to Load Yes
Size Range 100 to 1000 bp
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Product Line MassRuler
Quantity 2 x 500 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.