missing translation for 'onlineSavingsMsg'
Learn More

Atago™ MASTER-SUR/NM Model Clinical Refractometer

Brand:  Atago™ 2773

Additional Details : Weight : 0.10000kg

Product Code. 11456168

  • 335.00€ / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Clinical Refractometer
1.3330 to 1.3660
Urine S.G., Serum protein, Refractive index
110 g
1.000 to 1.060 Urine Specific Gravity, 0 to 12 g/100 mL Serum Protein
MASTER-SUR/NM
Designed for clinical purposes: Serum Protein, Specific Gravity of Urine and Refractive Index
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions