Learn More
Eppendorf™ Mastercycler X50p 96-Well High Pressure Thermal Cycler
Eppendorf™ Mastercycler X50 thermal cycler is the elegant synthesis of speed, flexibility and PCR optimization for research PCR applications and standardization for routine PCR applications. A new highly intuitive touch screen concept puts all of those benefits at your fingertips.
Brand:  Eppendorf™ 6315000015
Regulatory: This product is under eco-contribution tax.
Product Code. 15816731
  • 12515.00€ / Pack of 1
In Stock
Description
  • Heating rate: 5°C/s
  • Innovative 2D-Gradient for advanced PCR optimization
  • Heating rate: 5°C/s
  • Block temperature accuracy: ±0.15°C
  • Touch screen interface
  • Unit can control up to 9 other units (connected cyclers do not have a touch screen interface)
  • High pressure Lid
  • Small footprint
  • 2 year warranty
Certifications
Declaration of Conformity
Services
2 year warranty