Learn More
Masterflex™ Masterflex™ Polypropylene Sanitary Barbed Adapter
Product Code. 11891101
Change view
Click to view available options
For Use With:
3/4 in. Clamp Size x 1/2 in. Tubing I.D.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11891101 3/4 in. Clamp Size x 1/2 in. Tubing I.D. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11891101 Supplier Masterflex™ Supplier No. 3180527
Only null left
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With 3/4 in. Clamp Size x 1/2 in. Tubing I.D.
Min. Operating Temperature (Metric) -9°C
Packaging Quantity 1 Pack
Max. Operating Temperature (Metric) 82°C
Product Type Sanitary Barbed Adapter