missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Norprene™ Food A-60-F Spooled Precision Pump Tubing

Similar to Norprene™ but with FDA approval

Brand:  Masterflex™ 06415-14

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11806773

  • 915.00€ / Each
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 16
  • Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils
  • Heat, ozone, and UV light resistant, nonaging, nonoxidizing, superior acid and alkali resistance, opaque beige
  • Ideal for the food, dairy, and beverage industries
Specifications

Specifications

Autoclavable
Norprene™ Food (A 60 F)
Food and dairy applications
14 (1.6 mm)
1/16 in.
2.7 bar
132°C
1.6 mm
15,240 cm
-51°C
0.21 to 130 mL/min.
8.8 m
660 mm Hg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions