Learn More
Masterflex™ L/S™ Easy-Load™ Pump Head Mounting Hardware
Click to view available options
For Use With:
1 Pump Head
2 Pump Heads
3 Pump Heads
4 Pump Heads
Unit Size:
Each
Pack of 4
Description
Easy stack multiple pump heads on your Masterflex™ L/S™ drive
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
- Suitable for use with one Easy-Load™ pump head
Specifications
Specifications
| For Use With (Application) | Mounting L/S Easy-Load Pump Heads |
| For Use With | 3 Pump Heads |
| Material | Stainless Steel |
| Quantity | 1/Ea. |
| Type | Pump Head Mounting Hardware |