Learn More
Thermo Scientific™ Matrix™ Filtered Pipette Tips
Eliminate pipetter barrel contamination with these filtered pipette tips.
Brand:  Thermo Scientific™ 7325
Additional Details : Weight : 1.53000kg
 View more versions of this product
Product Code. 10059692
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Eliminate pipetter barrel contamination with Thermo Scientific™ Matrix™ Filtered Pipette Tips. Rigorously tested to ensure optimum performance, these filtered tips eliminate the risk of cross-contamination from aerosols created while pipetting genomic material, bacterial samples or other precious samples.
Features:
  • Stringent testing guarantees filter tips that effectively block aerosols
  • Hydrophobic polyethylene filter material ensures no particulates contaminate samples
  • Inadvertent contact with filter will not contaminate sample or result in sample loss
  • Easy-to-open packaging maintains product sterility while providing easy access
  • Hinged lid allows rack to be opened without laying lid on benchtop, helping maintain sterility
  • Racks are sealed in thick polyethylene bag for sterility, which is easy to open with simple snap-and-tear packaging
Stringent Filter Testing:
  • Filter must meet certain porosity requirements and must form a complete barrier across face of tip.
  • To ensure defects are not present, two specific tests are performed: mercury intrusion and particle count testing.
  • Mercury Intrusion Testing (QC 13.1 - QC 13.8) ensures the filter itself meets specific aerosol blocking requirements.
  • Particle Count Testing (Test Protocol QAS-0100) ensures filter is effective at blocking aerosols after installation into tip, also ensures proper shape and fit of filter.
  • Every lot of tips is tested by drawing a vacuum of aerosols against the filter and ensuring particles do not pass through.
Recommended for:
  • Bacteriology
  • Gel loading
  • PCR setup
  • Preventing cross-contamination during routine pipetting
Specifications
Specifications
250 μL
Sterile
Gilson Pipetman P200, Thermo Scientific Matrix Impact, Thermo Scientific™ Matrix™ Impact™ 250 μL
Filtered
Rack
Matrix
Product Suggestions