missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier for AAS, Pd(NO3)2 2000 mg/L in HNO3 1%, Honeywell Fluka™

Product Code. 16624464 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity unitSize
16624464 50 mL 50mL
1 options
This item is not returnable. View return policy

Product Code. 16624464

Brand: Honeywell Fluka E460250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Matrix Modifier for AAS
Density 1.01 g/cm3
pH <2
Quantity 50 mL
Grade Standards and Modifiers for AAS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.