missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ MATRIX PCR Attachment
Shop All IKA Products
Click to view available options
Holds:
384-well PCR Plates
96-well PCR Plates
Unit Size:
Each
Description
The thermoshakers MATRIX Orbital Delta plus and MATRIX Delta plus automatically recognize, which attachment is connected to the device via an interface in the locking mechanism. The attachment can be changed quickly and easily with an ergonomic lever. Suitable for: MATRIX Orbital Delta plus MATRIX Delta plus
Specifications
Specifications
| Holds | 96-well PCR Plates |
| Tube Capacity | 0.2 mL |
| Product Type | PCR Attachment |
| Dimensions (L x W x H) | 150 x 141 x 42 mm |
| For Use With (Application) | Samples with even the smallest volume, such as blood samples, pharmaceutical agents, ELISA assays/DNA/RNA samples |
| For Use With (Equipment) | MATRIX Orbital Delta Plus, MATRIX Delta Plus |
| No. of Positions | 96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction