Learn More
BOLA Maxi Propeller Blades
Product Code. 10326152
Click to view available options
Dimensions (L x W x H):
140 x 20 x 4 mm
200 x 20 x 6 mm
200 x 26 x 6 mm
Unit Size:
Each
Product Code. 10326152
Brand: BOLA C44116
Only null left
Add to basket
Add to basket
Maxi Propeller Blades for stainless steel shafts jacketed with PTFE, glass shafts or metal shafts
  • These solid stirrer blades are made of PTFE and have a set of clamp screws made of a PTFE/PEEK compound.
  • The blades can be fixed tightly on BOLA Stirrer Shafts by means of the clamp screws. A spanner wrench is included for easy assembly.
Specifications
Dimensions (L x W x H) 200 x 26 x 6 mm
Coating PTFE