Learn More
BOLA Maxi Propeller Blades
Click to view available options
Dimensions (L x W x H):
140 x 20 x 4 mm
200 x 20 x 6 mm
200 x 26 x 6 mm
Unit Size:
Each
Description
- These solid stirrer blades are made of PTFE and have a set of clamp screws made of a PTFE/PEEK compound.
- The blades can be fixed tightly on BOLA Stirrer Shafts by means of the clamp screws. A spanner wrench is included for easy assembly.
Specifications
Specifications
| Dimensions (L x W x H) | 200 x 26 x 6 mm |
| Coating | PTFE |