Learn More
3M™ Maxim™ Ballistic Safety Spectacles 13298-00000M
Product Code. 15789657
Click to view available options
:
Lens Tint: Clear
Product Code. 15789657
Brand: 3M™ 7000030022
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
3M™ Maxim™ Ballistic Safety Spectacles are designed specifically to wear in combination with a helmet and or ear muffs.
  • DX coating provides enhanced anti-scratch and anti-fog protection
  • Spectacles with ventilation channel across the lens to minimize misting
  • Design provides excellent coverage and good field of vision
  • Pliable, adjustable nose-pads and a soft-touch brow guard
  • Temples feature an adjustable pantoscopic angle
  • Low-profile design increases compatibility with helmets and hearing protection
  • Weighs only 28 g
TRUSTED_SUSTAINABILITY
Specifications
Lens Type DX
Lens Coating Anti-fog, Scratch-resistant
Description Model 13298-00000M
Lens Tint Clear
Product Type Ballistic Spectacles