missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maximum Recovery Diluent (Dehydrated)

Product Code. p-200112442
Click to view available options
Quantity:
500 g
2.5 kg
5kg
Unit Size:
3kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10229682

Brand: Thermo Scientific™ CM0733B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent (Dehydrated).

Maximum Recovery Diluent combines the protective effect of peptone in the diluting solution1 with the osmotic support of physiological saline1,2. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2

Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.

  • Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
  • Reduces organism multiplication in the sample for at least one hour during the dilution stage.

The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, BSI, EP, IDF, ISO, NMKL
Description Mannitol Salt Agar
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 52.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.