missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MBL Oligomer Human ELISA Kit

Product Code. 17737527
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17737527 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17737527 Supplier Invitrogen™ Supplier No. BMS2329

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

ELISA

MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11226
Assay Range 40 to 0 ng/mL
Assay Sensitivity 6 pg/mL
Conjugate HRP
Product Type ELISA Kit
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4153
Gene Symbol MBL2
Interassay CV <10%
Intraassay CV <6%
Kit Contents Pre-coated 96 well plate, Standard (2 vials), HRP Detection Antibody, Assay buffer Concentrate, Wash buffer concnetrate, TMB, Stop solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Serum, 10 μL|Plasma, 10 μL
Storage Requirements 2°C to 8°C
Target MBL2
Test Time 1 hr 20 min
Total Assay Time 3 hr 15 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.