missing translation for 'onlineSavingsMsg'
Learn More

MCM7 Mouse, Clone: MCM7/1468, enQuireBio™

Mouse Monoclonal Antibody

199.00€ - 510.00€

Specifications

Antigen MCM7
Clone MCM7/1468
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15920458
View Documents Promotion Details
EnquireBio
4176-MSM3-P1
100 μg
510.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15910458
View Documents Promotion Details
EnquireBio
4176-MSM3-P0
20 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

MCM7
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
4176
Recombinant human MCM7 protein fragment (aa195-319)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
88kDa
MCM7/1468
Immunohistochemistry
Unconjugated
RUO
NP_005907
MCM7
IgG2b
Protein A/G Purified
Nuclear
Raji or Ramos cells. Tonsil
Videos
SDS
Documents

Documents

Product Certifications