Learn More
BrandTech™ VACUUBRAND™ MD 1 Model Diaphragm Pump
GREENER_CHOICE
Product Code. 10678951
Click to view available options
Plug Type:
CEE
Voltage:
200/230 V
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10678951
Brand: BrandTech™ VACUUBRAND™ 20696080
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
An excellent choice for oil-free evacuation and pumping of non-corrosive gases down to 1.5 mbar ultimate vacuum.
  • Three-stage design with precisely guided flat diaphragms provides outstanding pumping speed even close to the ultimate vacuum, especially when compared with similarly rated two-stage diaphragm pumps
  • Ultra-low vibration with very long diaphragm lifetimes and stable ultimate vacuum documented with years of testing
  • Flow path consisting of aluminum and selected plastics (diaphragms and valves made of PTFE/FKM and FKM, respectively) the pumps are suitable for a wide range of applications with non-corrosive gases
  • Contamination-free pumping and evacuation
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 1.1 torr
Horsepower 0.11 hp
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance ATEX Conformity II 3G IIC T3 X, CAN/CSA C22.2 61010-1-12, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV, UL 61010A 61010-1:2012
Electrical Requirements 200/230 V 50/60 Hz 0.08 kw
Dimensions (L x W x H) 303 x 143 x 163 mm
Material Fluoroplastic (Case), Aluminum, Plastics, PTFE/FKM (Diaphragm), FKM (Valves)
Model MD 1
Noise Level 40 dbA
Plug Type CEE
Wattage 0.08 kw
Weight (Metric) 6.5 kg
Temperature (English) Max. 104°F
Flow Rate 0.7/0.8 CFM
Frequency 50/60 Hz
Overall Height (Metric) 215.9 mm
Overall Length (Metric) 302.26 mm
Overall Width (Metric) 142.24 mm
Pump Type Diaphragm Pump
Enclosure Rating IP42
No. of Pump Heads 4
Resistance Properties Chemical-Resistant
Temperature (English) Min 50°F
Applications Non-Corrosive Gases
Oil Type Oil-Free
No. of Stages 3
Maximum Pressure 16 psi
Voltage 200/230 V