Learn More
BrandTech™ VACUUBRAND™ ME 4C NT Dry Chemistry Diaphragm Pump
GREENER_CHOICE
Product Code. 10725774
Change view
Click to view available options
Plug Type:
CEE
Electrical Requirements:
230 V 50/60 Hz
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Electrical Requirements unitSize
10725774 CEE 230 V 50/60 Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10725774 Supplier BrandTech™ VACUUBRAND™ Supplier No. 20731200
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Suitable for laboratory tasks such as vacuum filtration, degassing, solid phase extraction and many others
  • Engineered to provide 40% higher flowrates at working vacuum level than other competitive pumps with similar free air capacity ratings
  • Whisper quite operation at ultimate vacuum
  • Less vibration than previous model
  • Flowpaths constructed of materials with excellent corrosion resistance (such as PTFE heads and diaphragms
  • Eliminates the misting, handling, and disposal of contaminated oil associated with rotary vane pumps Service intervals: 10,000 to 15,000 hours
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 52.5 torr
Horsepower 0.24 hp
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance ATEX Conformity II 3G IIC T3 X, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV
Electrical Requirements 230 V 50/60 Hz
Material PTFE (Pump Head), Fluoroplastic (Case)
Model ME 4C NT
Noise Level 45 dbA
Plug Type CEE
Weight (English) 24.47 lb.
Weight (Metric) 11.1 kg
Temperature (English) Max. 104°F
Flow Rate 2.3/2.6 CFM
Frequency 50/60 Hz
Overall Height (English) 7.79 in.
Overall Height (Metric) 198.12 mm
Overall Length (English) 10 in.
Overall Length (Metric) 254 mm
Overall Width (English) 9.56 in.
Overall Width (Metric) 24.28 cm
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 2
Resistance Properties Chemical-Resistant
Temperature (English) Min 50°F
Applications Filtration and Concentration of Solvents with Low Boiling Point
Oil Type Oil-Free
No. of Stages 1
Maximum Pressure 16 psi
Voltage 230 V