Learn More
Huenersdorf™ Measuring Cup
Click to view available options
Capacity (Metric):
250 mL
Description
- Closed handle
- Transparent
- 5 mL scale increments
Specifications
Capacity (Metric) | 250 mL |
Color | Transparent |
Material | Polypropylene |
Graduated | Yes |