missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Mechanical Stage for FBGG/2111/2112 Stands
Manipulate samples easier and with more comfort. Motic™ Mecanical Stagefor FBGG/2111/2112 Stands includes bidirectional movement.
Brand: Motic™ 1101000600552
Additional Details : Weight : 1.00000kg
Description
- Attachable x/y-stage
- 76×54mm travel range
- Stage for FBGG/2111/2112 stand
- Bidirectional movement
- Rotatable insert is available as an option
Order Info
dimProductType
Specifications
Mechanical Stage | |
150 x 150 mm | |
Black/White plate |
76 x 54mm travel range | |
FBGG/2111/2112 Stand | |
1 Ea. |