Learn More
Liebherr™ Pharmacy Fridge
Shop All Liebherr Products
Click to view available options
Capacity (Metric):
142 L
152 L
281 L
361 L
386 L
Door Style:
Glass
Self-closing
Solid
missing translation for 'unitSize'
Each
Beschreibung
- DIN 58345 compliant, width 600mm.
- Temperature set to +5°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
- Climate class 4, refrigerant R 600a.
- Digital controller with 0,1°C resolution, password keypad lock, RS 485 interface and volt-free alarm contact
- Access port for external temperature probe, additional safety thermostat
- Alarm log of last 3 events with min/max temperatures, duration etc.
- Integrated battery for continuous alarm and monitoring for min. 12 hrs in case of power failure
- Door lock, reversible hinges, self-closing door
- 45 kg shelf load
Spezifikation
Spezifikation
| Description | Pharmacy refrigerator |
| Capacity (Metric) | 281 L |
| Temperature Range | 5°C |
| Temperature Sensor | Pt100 |
| Refrigerant | R600A |
| Door Style | Self-closing |
| Door Material | Steel |
| Shelves | Plastic-coated Grid |
| Interior | White Plastic Containers |
| Certifications/Compliance | DIN 58345 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur