missing translation for 'onlineSavingsMsg'
Learn More

Mefenamic acid, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J62705.14

Quantity 25g

 View more versions of this product

Product Code. 15415029

  • 48.10€ / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

61-68-7
241.29
HYYBABOKPJLUIN-UHFFFAOYSA-N
4044
2-(2,3-dimethylanilino)benzoic acid
C15H15NO2
MFCD00051721
mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
CHEBI:6717
CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C

Specifications

61-68-7
MFCD00051721
14,5798
It is soluble in acetone,chloroform,dichloromethane,methanol. Insoluble in water.
CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C
241.29
CHEBI:6717
98%
C15H15NO2
25g
mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
HYYBABOKPJLUIN-UHFFFAOYSA-N
2-(2,3-dimethylanilino)benzoic acid
4044
241.29
Mefenamic acid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.