missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX MEK1 Protein, Active

Product Code. 17300174
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17300174

Brand: Eurofins DiscoverX 14429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal GST and C-terminal His6-tagged, recombinant full-length human MEK1, for use in Kinase Assays.

ERKs (extracellular signal-regulated kinase) are activated by the MEK (mitogen-activated protein kinase, or ERK kinase). MEKs are activated by members of the Raf family. The signaling pathway comprising Raf and ERK (extracellular signal-regulated kinase) lies downstream of the small guanine nucleotide binding protein Ras and mediates several apparently conflicting cellular responses, such as proliferation, apoptosis, growth arrest, differentiation, and senescence, depending on the duration and strength of the external stimulus and on cell type. An inhibitor assay may be of benefit for the treatment of cancer, diabetes, neurodegenerative disorders or asthma.

EMBL Z30163, possessing amino acid substitution A327G. Please note, introduction of the A327G mutation to the rabbit sequence (Z30163) results in expression of recombinant MEK1 protein that is 100% identical to wild-type human MEK1 described in SwissProt Q02750, note this SwissProt entry fails to record M1 and starts at P2.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 71 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST and C-terminal 6His tags
Expression System Recombinant enzyme expressed in E. coli cells
Protein Form Full-length
Purity or Quality Grade ≥72%
Protein MEK1, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.