Learn More
BrandTech™ VITLAB™ Melamine Formaldehyde Instrument Tray
Click to view available options
Dimensions (L x W x H):
290 x 160 x 35 mm
355 x 240 x 17 mm
428 x 288 x 17 mm
340 x 245 x 100 mm
Unit Size:
Pack of 5
Description
- White Melamine
- Flat shape
- Rounded corners
- Smooth surface
- Easy to clean
Specifications
Specifications
| Dimensions (L x W) | 355 x 240 mm |
| Dimensions (L x W x H) | 355 x 240 x 17 mm |
| Height (Metric) | 17 mm |
| Length (Metric) | 355 mm |
| Material | Melamine Formaldehyde |
| Type | Instrument Tray |
| Width (Metric) | 240 mm |
| Quantity | 1 Each |
| Shape | Flat |