Learn More
B Braun™ Surface Spray Disinfectant
Use B Braun™ Surface Spray Disinfectant for rapid surface disinfection with Rapid Meliseptrol. The broad-spectrum disinfectant has a 5-minute reaction time.
Brand: B Braun™ 18567
Additional Details : CAS : 71-23-8 Weight : 4.55000kg Transport : UN number : 1274 Chem class : 3 Pack group : III
Description
- Rapid Meliseptrol distribution
- Free of CFC's
- For small surfaces and equipment
- Colorless
Specifications
Disinfectant | |
Colorless | |
Canister | |
5 L |
∼7 | |
89°C | |
For small surfaces and equipment |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : ethanol, butanone