missing translation for 'onlineSavingsMsg'
Learn More

Serva Electrophoresis™ Membra-Cel™ Dialysis Tubing

Product Code. 11722597
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 m
5 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11722597 5 m Each
11732597 30 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11722597 Supplier Serva Electrophoresis™ Supplier No. 4431001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Dialysis Tubing
MWCO 3.5 kDa
Material Cellulose
Diameter (Metric) 16 mm
Length (Metric) 5 m
Capacity (Metric) 2 mL/cm
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.