missing translation for 'onlineSavingsMsg'
Learn More

Serva Electrophoresis™ Membra-Cel™ Dialysis Tubing

Product Code. 11732597
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 m
5 m
Unit Size:
Each
Product Code. Length (Metric) unitSize
11732597 30 m Each
11722597 5 m Each
2 options
This item is not returnable. View return policy

Product Code. 11732597

Brand: Serva Electrophoresis™ 4431002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Dialysis Tubing
MWCO 3.5 kDa
Material Cellulose
Diameter (Metric) 16 mm
Length (Metric) 30 m
Capacity (Metric) 2 mL/cm
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.