Learn More
Serva Electrophoresis™ Membra-Cel™ Dialysis Tubing
Product Code. p-8010998
Encompass_Preferred
Click to view available options
Length (Metric):
30 m
5 m
Unit Size:
Each
Product Code. 11722597
Brand: Serva Electrophoresis™ 4431001
Specifications
Product Type Dialysis Tubing
MWCO 3.5 kDa
Material Cellulose
Diameter (Metric) 16 mm
Length (Metric) 5 m
Capacity (Metric) 2 mL/cm
Quantity 1 Each