Learn More
Emma Safety Footwear Mento Safety Shoes
Water repellant for up to 4 hours and ideally suited to working outdoors. Designed to offer exceptional all over foot protection with a reinforced steel plate in the sole, and strong, supportive leather uppers. A double-density polyurethane sole protects feet from dangers above and below.
Brand: Emma Safety Footwear MM58154839
Additional Details : Weight : 1.97200kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Easy Twist Outsole
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
EN ISO 20345:2011 | |
Mento | |
Leather | |
Steel |
Brown | |
Hydro-Tec™ Sanitized Silver | |
39 |