missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Product Code. p-7050297
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11317888

Brand: Thermo Scientific Alfa Aesar 036419.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Mercury(I) chloride is used in medicine as a diuretic and purgative, uses in cosmetics as soaps and skin lightening creams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.084
MDL Number MFCD00011043
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]

Specifications

Melting Point 400°C (sublimation)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Hg2Cl2
UN Number UN2025
Merck Index 14,5894
Solubility Information Soluble in (0.002g/L )water.
Formula Weight 472.09
Percent Purity ≤99.5%
Grade ACS
Odor Odorless
Sensitivity Light and moisture sensitive
Density 7.15 g/mL
Chemical Name or Material Mercury(I) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.