missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MesenCult™ Expansion Kit (Mouse)
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- MesenCult™ Expansion Kit (Mouse) is standardized for the culture of mouse mesenchymal stromal cells (MSCs; also known as mesenchymal stem cells) and mouse embryonic fibroblasts (MEFs)
- The kit includes MesenCult™ Basal Medium (Mouse), MesenCult™ 10X Supplement (Mouse), and MesenPure™
- MesenCult™ Expansion Medium has been optimized for the derivation and expansion of mouse MSCs and MEFs in vitro as well as for the detection of colony-forming unit–fibroblasts (CFU-F)
- This kit was optimized using cells from the mouse strain C57BL/6
- To facilitate the enrichment of MSCs and MEFs during cell culture without serial passaging and frequent medium changes, simply add MesenPure™ to complete MesenCult™ Expansion Medium just prior to use
- NOTE: MesenCult™ Expansion Medium must be supplemented with L-Glutamine (Catalog #07100)
Specifications
Specifications
| Cell Type | Mesenchymal Stem and Progenitor Cells, Mouse Embryonic Fibroblasts |
| For Use With (Application) | Cell Culture, Colony Assay, Expansion |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction