missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MesenCult™ MSC Stimulatory Supplement (Human)
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
- MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is a standardized, serum-containing supplement for the culture of human mesenchymal stem cells (MSCs)
- MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is optimized for the expansion of human mesenchymal stem cells in vitro as well as their enumeration using the colony-forming unit - fibroblast (CFU-F) assay when combined with MesenCult™ MSC Basal Medium (Human)
- Components are pre-screened to minimize lot-to-lot variability
- This supplement is also a component of MesenCult™ Proliferation Kit (Human; Catalog #05411)
Specifications
Specifications
| Cell Type | Mesenchymal Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Expansion |
| Quantity | 50 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction