missing translation for 'onlineSavingsMsg'
Learn More

Isodrin in Methanol 10μg/mL, Fisher Chemical™

Catalog Number 12949843
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10μg/mL
CAS 465-73-6
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C12H8Cl6
InChI Key QBYJBZPUGVGKQQ-HSECHKQMSA-N
SMILES C1C2C=CC1C3C2C4(C(=C(C3(C4(Cl)Cl)Cl)Cl)Cl)Cl
Molecular Weight (g/mol) 364.896
PubChem CID 102090390
Solvent or Matrix Methanol
Chemical Name or Material Isodrin
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'