missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Terphenyl D14 in DCM 500μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 1718-51-0 |
| Molecular Formula | C18H14 |
| Concentration | 500 μg/mL |
| InChI Key | XJKSTNDFUHDPQJ-WZAAGXFHSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3 |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene |
| Solvent or Matrix | Dichloromethane |
| Molecular Weight (g/mol) | 244.395 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction