missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paraquat dichloride in Water 10μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 10mu;g/mL |
CAS | 1910-42-5 |
Packaging | Ampoule(s) |
Quantity | 10 mL |
Molecular Formula | C12H14Cl2N2 |
MDL Number | MFCD00150001 |
InChI Key | FIKAKWIAUPDISJ-UHFFFAOYSA-L |
SMILES | [Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1 |
IUPAC Name | 1,1'-dimethyl-[4,4'-bipyridine]-1,1'-diium dichloride |
Molecular Weight (g/mol) | 257.16 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'