missing translation for 'onlineSavingsMsg'
Learn More

Terbuthylazine-2-hydroxy in Acetone 10μg/mL, Fisher Chemical™

Catalog Number 12970124
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10μg/mL
CAS 66753-07-9
Packaging Ampoule
Quantity 10 mL
Type Organic Single-component Standard Solution
Molecular Formula C9H17N5O
UN Number 1090
InChI Key OYTCZOJKXCTBHG-UHFFFAOYSA-N
SMILES CCNC1=NC(=O)N=C(N1)NC(C)(C)C
IUPAC Name 2-(tert-butylamino)-6-(ethylamino)-1H-1,3,5-triazin-4-one
Molecular Weight (g/mol) 211.269
PubChem CID 5151089
ChEBI CHEBI:83471
Solvent or Matrix Acetone
Physical Form Liquid
Chemical Name or Material Terbuthylazine-2-hydroxy
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Terbuthylazine-2-hydroxy 10ug/ml in acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'