Learn More
Scientific Industries™ Metal Clip Plates for Volumetric Flasks
Securely holds up to four 100 mL volumetric flasks.
Brand:  Scientific Industries™ SI-1136
Additional Details : Weight : 0.25000kg
Product Code. 10165692
  • 49.70€ / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
Metal clip plate to accommodate up to four 100 mL volumetric flasks. This can be attached to the magnetic platform for use with the Roto-Shake Genie, Rotator Genie, Enviro-Genie or Incubator-Genie.